Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Oxyphencyclimine hydrochloride | 125-52-0 | 99.6% | 50 MG
SDP

Catalog No. 5000408832
Encompass_Preferred

Oxyphencyclimine hydrochloride is a new tertiary amine that has been investigated for its effects on the peripheral parasympathetic system. It is provided as a white to off-white solid and is soluble in DMSO at concentrations of ≥ 30 mg/mL. This product is intended for research use only.

  • Investigated for effects on the peripheral parasympathetic system
  • Molecular weight: 380.91
  • Formula: C20H29ClN2O3
  • Appearance: White to off-white solid
  • Solubility: Soluble in DMSO at concentrations of ≥ 30 mg/mL
  • Storage (powder): 4°C, sealed, away from moisture and light
  • Storage (in solvent): -80°C for 6 months; -20°C for 1 month
  • Related signaling pathways include GPCR/G protein, neuronal signaling, and mAChR
  • Classified as a muscarinic acetylcholine receptor (mAChR) antagonist

Catalog No. 50-004-08832 Supplier Medchemexpress LLC Supplier No. HYB095450MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Oxyphencyclimine hydrochloride is a new tertiary amine that has been investigated for its effects on the peripheral parasympathetic system. It is provided as a white to off-white solid and is soluble in DMSO at concentrations of ≥ 30 mg/mL. This product is intended for research use only.

  • Investigated for effects on the peripheral parasympathetic system
  • Molecular weight: 380.91
  • Formula: C20H29ClN2O3
  • Appearance: White to off-white solid
  • Solubility: Soluble in DMSO at concentrations of ≥ 30 mg/mL
  • Storage (powder): 4°C, sealed, away from moisture and light
  • Storage (in solvent): -80°C for 6 months; -20°C for 1 month
  • Related signaling pathways include GPCR/G protein, neuronal signaling, and mAChR
  • Classified as a muscarinic acetylcholine receptor (mAChR) antagonist

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.