Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Oxyphenisatin acetate | 115-33-3 | MFCD00022793 | 98.0% | 401.41 | C24H19NO5 | 50 MG
SDP

Catalog No. 5000246842
Encompass_Preferred

Oxyphenisatin acetate is the acetylated prodrug of oxyphenisatin, historically used as a laxative. This listing describes a research-grade chemical supplied for analytical and laboratory studies; it is intended for research use only and not for human or veterinary use.

  • Acetylated prodrug of oxyphenisatin.
  • Used in biochemical and autophagy-related research.
  • CAS number: 115-33-3.
  • Molecular formula C24H19NO5 and molecular weight ~401.41.
  • Typically supplied as a solid with high purity (commonly ≥98%).

Catalog No. 50-002-46842 Supplier Medchemexpress LLC Supplier No. HY10171450MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Oxyphenisatin acetate is the acetylated prodrug of oxyphenisatin, historically used as a laxative. This listing describes a research-grade chemical supplied for analytical and laboratory studies; it is intended for research use only and not for human or veterinary use.

  • Acetylated prodrug of oxyphenisatin.
  • Used in biochemical and autophagy-related research.
  • CAS number: 115-33-3.
  • Molecular formula C24H19NO5 and molecular weight ~401.41.
  • Typically supplied as a solid with high purity (commonly ≥98%).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.