Learn More
Medchemexpress LLC Oxyphenisatin acetate | 115-33-3 | MFCD00022793 | 98.3% | 401.41 | C24H19NO5 | 5 MG

Supplier: Medchemexpress LLC HY1017145MG
Oxyphenisatin acetate is the acetylated pro-drug of oxyphenisatin, historically used as a laxative. It is provided as a research-grade chemical/analytical standard for laboratory and analytical applications and is not intended for human or clinical use.
- Purity typically 98.3% (batch dependent).
- CAS number 115-33-3.
- Molecular formula C24H19NO5 and molecular weight ~401.41.
- Intended for research and analytical applications only.
- Available in small quantity formats for analytical use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.