Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Oxyphenonium bromide | 50-10-2 | 99.4% | 428.40 g/mol | C21H34BrNO3 | 10 MG
SDP

Catalog No. 5000298899
Encompass_Preferred

Oxyphenonium bromide is an antimuscarinic research reagent that acts as a muscarinic acetylcholine receptor (mAChR) antagonist. It is supplied as the bromide salt for pharmacology and respiratory physiology studies, including assays of bronchoconstriction and receptor modulation. Intended for research and analytical use.

  • Antimuscarinic agent, mAChR antagonist
  • High purity (reported 99.39%)
  • CAS number 50-10-2 for unambiguous identification
  • Molecular formula C21H34BrNO3 and molecular weight 428.40 g/mol
  • Provided as a 10 mg research sample
  • Intended for research and analytical applications; not for human therapeutic use

Catalog No. 50-002-98899 Supplier Medchemexpress LLC Supplier No. HYB1719A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Oxyphenonium bromide is an antimuscarinic research reagent that acts as a muscarinic acetylcholine receptor (mAChR) antagonist. It is supplied as the bromide salt for pharmacology and respiratory physiology studies, including assays of bronchoconstriction and receptor modulation. Intended for research and analytical use.

  • Antimuscarinic agent, mAChR antagonist
  • High purity (reported 99.39%)
  • CAS number 50-10-2 for unambiguous identification
  • Molecular formula C21H34BrNO3 and molecular weight 428.40 g/mol
  • Provided as a 10 mg research sample
  • Intended for research and analytical applications; not for human therapeutic use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.