Learn More
Broadley James Corporation D145 12 mm OxyProbe with Right Angle Connector, 220mm Sensor
Catalog No. NC1829713
Encompass
The OxyProbe 12 mm dissolved oxygen sensor with right angled connector offers the most rugged and versatile design for benchtop vessels. The compact profile saves space on a crowded headplate. It will fit directly into any 12 mm multipurpose port with standard compression fittings or Pg13.5 threads. The small diameter facilitates DO measurement in small volumes, especially bottles and spinner flasks.
Catalog No. NC1829713 Supplier Broadley James Corporation Supplier No. D145B220PTD9
The OxyProbe 12 mm dissolved oxygen sensor with right angled connector offers the most rugged and versatile design for benchtop vessels. The compact profile saves space on a crowded headplate. It will fit directly into any 12 mm multipurpose port with standard compression fittings or Pg13.5 threads. The small diameter facilitates DO measurement in small volumes, especially bottles and spinner flasks.