Learn More
Bellco Glass, Inc. OxyProbeモ DO Probe,12x120mm Broadley-James
SDP
Catalog No. 501946306
Encompass_Preferred
The OxyProbe 12mm dissolved oxygen sensors offer the most rugged and versatile design for benchtop vessels. The compact profile saves space and will fit directly into any port with standard compression fittings or threads. The small diameter facilitates DO measurement in small volumes such as spinner flasks and bottles. The polarographic sensor utilizes a long silver anode and small cathode (per the industry standard), it also uses the same gas permeable silicone PTFEcomposite membrane. Reinforced with stainless steel mesh the OxyProbe withstands steam in place and caustic clean in place applications. FDA compliant O-Rings are on all product contact surfaces.
Catalog No. 50-194-6306 Supplier Bellco Glass, Inc. Supplier No. 780331330
The OxyProbe 12mm dissolved oxygen sensors offer the most rugged and versatile design for benchtop vessels. The compact profile saves space and will fit directly into any port with standard compression fittings or threads. The small diameter facilitates DO measurement in small volumes such as spinner flasks and bottles. The polarographic sensor utilizes a long silver anode and small cathode (per the industry standard), it also uses the same gas permeable silicone PTFEcomposite membrane. Reinforced with stainless steel mesh the OxyProbe withstands steam in place and caustic clean in place applications. FDA compliant O-Rings are on all product contact surfaces.