Learn More
Broadley James Corporation OXYPROBE ELECTROLYTE 25ML
Catalog No. NC9053568
Encompass
In critical biotech applications the electrolyte should be changed after every run to yield accurate results. The shelf life of this solution is approximately 2 years. Lot numbers and expiration dates are clearly marked on each bottle for tracking.
Catalog No. NC9053568 Supplier Broadley James Corporation Supplier No. AS3140C300025
In critical biotech applications the electrolyte should be changed after every run to yield accurate results. The shelf life of this solution is approximately 2 years. Lot numbers and expiration dates are clearly marked on each bottle for tracking.