Learn More
Medchemexpress LLC HY-19657 50mg Medchemexpress, Oxypurinol CAS:2465-59-0 Purity:>98%
SDP
Catalog No. 501970028
Encompass_Preferred
Medchemexpress, HY-19657 50mg Oxypurinol CAS:2465-59-0 Oxipurinol (Oxipurinol), the major active metabolite of Allopurinol, is an inhibitor of xanthine oxidase. Oxipurinol can be used to regulate blood urate levels and treat gout. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0028 Supplier Medchemexpress LLC Supplier No. HY1965750MG
Medchemexpress, HY-19657 50mg Oxypurinol CAS:2465-59-0 Oxipurinol (Oxipurinol), the major active metabolite of Allopurinol, is an inhibitor of xanthine oxidase. Oxipurinol can be used to regulate blood urate levels and treat gout. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.