Learn More
Oxyrase Oxypras Bbe/Kvl Bi-Plate 10Pk
SDP
Catalog No. NC9743189
Encompass
For large volume customers. 10-packs are 10 sterile BBE/KVL Bi-Plate PRAS plates aseptically packaged into a single bag for customers that don't have room or time for individually packaged plates.
Catalog No. NC9743189 Supplier Oxyrase Supplier No. BBCD10
For large volume customers. 10-packs are 10 sterile BBE/KVL Bi-Plate PRAS plates aseptically packaged into a single bag for customers that don't have room or time for individually packaged plates.