Learn More
Oxyrase Oxyrase For Agar 100Ml Bottle 1/Pk
SDP
Catalog No. NC9224790
Encompass
Grow and isolate anaerobes. Produce anaerobic conditions in custom agar. Saves time, money and increases efficiency. Eliminates the need for bags, jars and chambers. Best results when utilized in conjunction with OxyDishes.
This item is not returnable. View return policy
Catalog No. NC9224790 Supplier Oxyrase Supplier No. OA0100
Grow and isolate anaerobes. Produce anaerobic conditions in custom agar. Saves time, money and increases efficiency. Eliminates the need for bags, jars and chambers. Best results when utilized in conjunction with OxyDishes.