Learn More
Oxyrase Oxyrase for Agar, 1x50mL
SDP
Supplier:  Oxyrase OA0050
Encompass
Grow and isolate anaerobes. Produce anaerobic conditions in custom agar. Saves time, money and increases efficiency. Eliminates the need for bags, jars and chambers. Best results when utilized in conjunction with OxyDishes.
Catalog No. 50-300-2021