Learn More
Oxyrase Oxyrase OxyPRAS StrictAna Blood Agar Plate
SDP
Catalog No. NC3813077
Encompass
Microbiology Media, Plated Media, Selective Media, Oxyrase, OxyPRAS StrictAna BAP, Isolate obligate anaerobes from polymicrobial samples, Inhibition of common facultatives by action of Gentamycin, Prevents overgrowth of fast growing and motile facultatives, Brucella Blood agar supplemented with Hemin, Vit K, and Gentamycin, anaeobic specimen primary set up, Consists of 10 individually packaged plates, 3 month shelf life.
Catalog No. NC3813077 Supplier Oxyrase Supplier No. PSANABA
Microbiology Media, Plated Media, Selective Media, Oxyrase, OxyPRAS StrictAna BAP, Isolate obligate anaerobes from polymicrobial samples, Inhibition of common facultatives by action of Gentamycin, Prevents overgrowth of fast growing and motile facultatives, Brucella Blood agar supplemented with Hemin, Vit K, and Gentamycin, anaeobic specimen primary set up, Consists of 10 individually packaged plates, 3 month shelf life.