Learn More
Oxyrase Ec-Oxyrase Enzyme System, Sterile, 50Ml Bottle 1/Pk
SDP
Numéro de catalogue. NC9176863
Encompass
Oxyrase Unit defined as amount of activity that reduces dissolved oxygen @ 37°C & pH 8.4 in a phosphate buffer with lactate substrate @ rate of 1% per second. 0.3 unit of EC-Oxyrase reduces dissolved oxygen @ saturation to levels measured in parts per billion (ppb) in about 4 minutes. Active over wide temperature range. Operates over wide pH range 6.8 to 9.4.
Numéro de catalogue. NC9176863 Fournisseur Oxyrase Code fournisseur EC0050
Ajouter au panier
Ajouter au panier
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Oxyrase Unit defined as amount of activity that reduces dissolved oxygen @ 37°C & pH 8.4 in a phosphate buffer with lactate substrate @ rate of 1% per second. 0.3 unit of EC-Oxyrase reduces dissolved oxygen @ saturation to levels measured in parts per billion (ppb) in about 4 minutes. Active over wide temperature range. Operates over wide pH range 6.8 to 9.4.
missing translation for 'productTitle'
missing translation for 'pleaseSelectIssue'
missing translation for 'byClickingSubmit' missing translation for 'privacyPolicy'.