Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Oxyresveratrol 2-O-β-D-glucopyranoside | 392274-22-5 | 5 MG
SDP

Catalog No. 5000419545
Encompass_Preferred

Oxyresveratrol 2-O-β-D-glucopyranoside is a phenolic compound isolated from the root of *Morus nigra*. It functions as an effective tyrosinase inhibitor with an IC50 of 29.75 μM. This product is intended for research use only.

  • Phenolic compound isolated from *Morus nigra* root
  • Effective tyrosinase inhibitor
  • Intended for research use only
  • Purity: 98.93%
  • Molecular weight: 406.38
  • Chemical formula: C20H22O9
  • Appearance: White to off-white solid
  • Target: Tyrosinase

Catalog No. 50-004-19545 Supplier Medchemexpress LLC Supplier No. HYN35165MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Oxyresveratrol 2-O-β-D-glucopyranoside is a phenolic compound isolated from the root of *Morus nigra*. It functions as an effective tyrosinase inhibitor with an IC50 of 29.75 μM. This product is intended for research use only.

  • Phenolic compound isolated from *Morus nigra* root
  • Effective tyrosinase inhibitor
  • Intended for research use only
  • Purity: 98.93%
  • Molecular weight: 406.38
  • Chemical formula: C20H22O9
  • Appearance: White to off-white solid
  • Target: Tyrosinase

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.