Learn More
TARGETMOL CHEMICALS INC OXYRESVERATROL 500MG
SDP
Catalog No. 502580015
Encompass_Preferred
Also available in 50 mg 100 mg 1000 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. 1. Oxyresveratro can inhibit tyrosinase activity. 2. Oxyresveratro is useful to trauma models as toxicity to glia could be beneficial by inhibiting reactive gliosis. 3. Oxyresveratrol (trans-Oxyresveratrol) has antioxidant activity can reduce neuronal oxidative damage and protect hepatocytes against oxidative stress and mitochondrial dysfunction which may be associated with activation of Nrf2. purity: 99%
Catalog No. 50-258-0015 Supplier TARGETMOL CHEMICALS INC Supplier No. T3S1068500MG
Also available in 50 mg 100 mg 1000 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. 1. Oxyresveratro can inhibit tyrosinase activity. 2. Oxyresveratro is useful to trauma models as toxicity to glia could be beneficial by inhibiting reactive gliosis. 3. Oxyresveratrol (trans-Oxyresveratrol) has antioxidant activity can reduce neuronal oxidative damage and protect hepatocytes against oxidative stress and mitochondrial dysfunction which may be associated with activation of Nrf2. purity: 99%