Learn More
Cayman Chemical Oxyresveratrol, Purity: ≥98%, CAS: 29700-22-9, 50 mg
SDP
Catalog No. 502955600
Encompass_Preferred
A resveratrol analog; scavenges H2O2 and NO (IC50 = 45.3 μM) and DPPH (IC50 = 28.9 μM); reduces brain infarct volume and reduces cytochrome c release and caspase-3 activation in an in vivo model of stroke at 10 mg/kg; inhibits tyrosinase activity (IC50s = 1.2 and 52.7 μM in mushroom and mouse melanoma B-16 cells, respectively)
Catalog No. 50-295-5600 Supplier Cayman Chemical Supplier No. 1202850 MG
A resveratrol analog; scavenges H2O2 and NO (IC50 = 45.3 μM) and DPPH (IC50 = 28.9 μM); reduces brain infarct volume and reduces cytochrome c release and caspase-3 activation in an in vivo model of stroke at 10 mg/kg; inhibits tyrosinase activity (IC50s = 1.2 and 52.7 μM in mushroom and mouse melanoma B-16 cells, respectively)