Learn More
AdipoGen Oxyresveratrol
SDP
Catalog No. 502560339
Encompass_Preferred
Chemical. CAS 29700-22-9. Formula C14H12O4. MW 244.2. Synthetic. Naturallly occuring analog of resveratrol. Potent antioxidant and free-radical scavenger. Shown to have neuroprotective activity against cerebral ischemia and against traumatic injury. Apoptosis inhibitor in transient cerebral ischemia. Inhibits viral DNA replication and late viral protein synthesis of African Swine fever virus. Has depigmenting effects by effectively inhibiting tyrosinase activity, which catalyzes the rate-limiting step in synthesizing melanin pigments. Has anti-inflammatory properties based on inhibition of iNOS expression through down-regulation of NF-kB binding activity and significant inhibition of COX-2 activity. Antihyperlipidemic agent.
Catalog No. 50-256-0339 Supplier AdipoGen Supplier No. CDXO0035G001
Chemical. CAS 29700-22-9. Formula C14H12O4. MW 244.2. Synthetic. Naturallly occuring analog of resveratrol. Potent antioxidant and free-radical scavenger. Shown to have neuroprotective activity against cerebral ischemia and against traumatic injury. Apoptosis inhibitor in transient cerebral ischemia. Inhibits viral DNA replication and late viral protein synthesis of African Swine fever virus. Has depigmenting effects by effectively inhibiting tyrosinase activity, which catalyzes the rate-limiting step in synthesizing melanin pigments. Has anti-inflammatory properties based on inhibition of iNOS expression through down-regulation of NF-kB binding activity and significant inhibition of COX-2 activity. Antihyperlipidemic agent.