Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Oxysophocarpine | 26904-64-3 | MFCD08689951 | ≥98.0% | 262.35 | C15H22N2O2 | 10 MG
SDP

Catalog No. 5000295946
Encompass_Preferred

Oxysophocarpine is a quinolizidine alkaloid supplied as an analytical research standard for laboratory use. It is provided as a solid powder in small-mass vials and is intended for biochemical and pharmacological studies; store protected from light at low temperature.

  • Alkaloid research standard for biochemical and pharmacological studies.
  • Reported neuroprotective and anti-nociceptive activity.
  • Molecular formula C15H22N2O2; molecular weight 262.35.
  • Purity ≥98.0%.
  • Supplied in small-mass vials (1 mg-10 mg); store at 4°C protected from light; solutions require low-temperature storage.

Catalog No. 50-002-95946 Supplier Medchemexpress LLC Supplier No. HYN074610MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Oxysophocarpine is a quinolizidine alkaloid supplied as an analytical research standard for laboratory use. It is provided as a solid powder in small-mass vials and is intended for biochemical and pharmacological studies; store protected from light at low temperature.

  • Alkaloid research standard for biochemical and pharmacological studies.
  • Reported neuroprotective and anti-nociceptive activity.
  • Molecular formula C15H22N2O2; molecular weight 262.35.
  • Purity ≥98.0%.
  • Supplied in small-mass vials (1 mg-10 mg); store at 4°C protected from light; solutions require low-temperature storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.