Learn More
Medchemexpress LLC HY-N0746 5mg Medchemexpress, Oxysophocarpine CAS:26904-64-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN07465MG
Encompass_Preferred
Medchemexpress, HY-N0746 5mg Oxysophocarpine CAS:26904-64-3 Oxysophocarpine is an alkaloid extracted from Siphocampylus verticillatus. Oxysophocarpine has neuroprotective and anti-nociceptive effects on the central and peripheral nervous systems. Oxysophocarpine inhibits the growth and metastasis of  oral squamous cell carcinoma (OSCC). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2815