Learn More
Oxyrase Oxystasis Preservation Product, 10Ml Bottle 1/Pk
SDP
Catalog No. NC1274425
Encompass
Preserve and protect biological material at <-20°C. Quantitative recovery. Does not freeze. Easier and faster working with a liquid suspension. Saves time and money compared to cryopreservation.
This item is not returnable. View return policy
Catalog No. NC1274425 Supplier Oxyrase Supplier No. PROSTS
Preserve and protect biological material at <-20°C. Quantitative recovery. Does not freeze. Easier and faster working with a liquid suspension. Saves time and money compared to cryopreservation.