Learn More
TARGETMOL CHEMICALS INC Oxytetracycline 200MG
SDP
Catalog No. 502880509
Encompass_Preferred
Also available in 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Oxytetracycline (Terramycin) is a TETRACYCLINE analog isolated from the actinomycete STREPTOMYCES rimosus and used in a wide variety of clinical conditions. Purity 98.25%
Catalog No. 50-288-0509 Supplier TARGETMOL CHEMICALS INC Supplier No. T0895200MG
Also available in 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Oxytetracycline (Terramycin) is a TETRACYCLINE analog isolated from the actinomycete STREPTOMYCES rimosus and used in a wide variety of clinical conditions. Purity 98.25%