Learn More
Medchemexpress LLC Oxytetracycline | 79-57-2 | 99.1% | 1 G
SDP
Catalog No. 5000406074
Encompass_Preferred
Oxytetracycline is an antibiotic belonging to the tetracycline class. It potently inhibits Gram-negative and Gram-positive bacteria. Oxytetracycline is a protein synthesis inhibitor that prevents the binding from aminoacil-tRNA to the complex m-ribosomal RNA. It also possesses anti-HSV-1 activity.
  • Antibiotic (tetracycline class)
  • Potent inhibitor of Gram-negative and Gram-positive bacteria
  • Protein synthesis inhibitor
  • Possesses anti-HSV-1 activity
Catalog No. 50-004-06074 Supplier Medchemexpress LLC Supplier No. HYB02751G
Oxytetracycline is an antibiotic belonging to the tetracycline class. It potently inhibits Gram-negative and Gram-positive bacteria. Oxytetracycline is a protein synthesis inhibitor that prevents the binding from aminoacil-tRNA to the complex m-ribosomal RNA. It also possesses anti-HSV-1 activity.
  • Antibiotic (tetracycline class)
  • Potent inhibitor of Gram-negative and Gram-positive bacteria
  • Protein synthesis inhibitor
  • Possesses anti-HSV-1 activity