Learn More
Medchemexpress LLC Oxytetracycline | 79-57-2 | 99.2% | 5 G
SDP
Catalog No. 5000408747
Encompass_Preferred
Oxytetracycline is an antibiotic belonging to the tetracycline class. It potently inhibits Gram-negative and Gram-positive bacteria. This compound functions as a protein synthesis inhibitor and exhibits anti-HSV-1 activity.
  • Inhibits Gram-negative and Gram-positive bacteria
  • Acts as a protein synthesis inhibitor
  • Exhibits anti-HSV-1 activity
Catalog No. 50-004-08747 Supplier Medchemexpress LLC Supplier No. HYB02755G
Oxytetracycline is an antibiotic belonging to the tetracycline class. It potently inhibits Gram-negative and Gram-positive bacteria. This compound functions as a protein synthesis inhibitor and exhibits anti-HSV-1 activity.
  • Inhibits Gram-negative and Gram-positive bacteria
  • Acts as a protein synthesis inhibitor
  • Exhibits anti-HSV-1 activity