Learn More
Medchemexpress LLC Oxytetracycline | 79-57-2 | MFCD00003700 | 99.2% | 10 G
SDP
Catalog No. 5000406021
Encompass_Preferred
Oxytetracycline is an antibiotic from the tetracycline class. It potently inhibits Gram-negative and Gram-positive bacteria. This antibiotic functions as a protein synthesis inhibitor by preventing the binding of aminoacyl-tRNA to the ribosomal RNA complex. It also exhibits anti-HSV-1 activity.
  • Antibiotic from the tetracycline class
  • Potently inhibits Gram-negative and Gram-positive bacteria
  • Functions as a protein synthesis inhibitor
  • Prevents binding of aminoacyl-tRNA to ribosomal RNA complex
  • Exhibits anti-HSV-1 activity
  • Member of the bacterial aromatic polyketide family
  • Synthesized by a type II polyketide synthase
Catalog No. 50-004-06021 Supplier Medchemexpress LLC Supplier No. HYB027510G
Oxytetracycline is an antibiotic from the tetracycline class. It potently inhibits Gram-negative and Gram-positive bacteria. This antibiotic functions as a protein synthesis inhibitor by preventing the binding of aminoacyl-tRNA to the ribosomal RNA complex. It also exhibits anti-HSV-1 activity.
  • Antibiotic from the tetracycline class
  • Potently inhibits Gram-negative and Gram-positive bacteria
  • Functions as a protein synthesis inhibitor
  • Prevents binding of aminoacyl-tRNA to ribosomal RNA complex
  • Exhibits anti-HSV-1 activity
  • Member of the bacterial aromatic polyketide family
  • Synthesized by a type II polyketide synthase