Learn More
Medchemexpress LLC Oxytetracycline (dihydrate) | 6153-64-6 | 99.4% | 10 G
SDP
Catalog No. 5000406001
Encompass_Preferred
Oxytetracycline dihydrate is an antibiotic belonging to the tetracycline class. It potently inhibits both Gram-negative and Gram-positive bacteria by acting as a protein synthesis inhibitor, preventing the binding of aminoacyl-tRNA to the m-ribosomal RNA complex. This compound also exhibits anti-HSV-1 activity.
  • Potently inhibits Gram-negative and Gram-positive bacteria
  • Acts as a protein synthesis inhibitor
  • Exhibits anti-HSV-1 activity
  • Belongs to the tetracycline class of antibiotics
  • Classified as a phenol
Catalog No. 50-004-06001 Supplier Medchemexpress LLC Supplier No. HYB0275B10G
Oxytetracycline dihydrate is an antibiotic belonging to the tetracycline class. It potently inhibits both Gram-negative and Gram-positive bacteria by acting as a protein synthesis inhibitor, preventing the binding of aminoacyl-tRNA to the m-ribosomal RNA complex. This compound also exhibits anti-HSV-1 activity.
  • Potently inhibits Gram-negative and Gram-positive bacteria
  • Acts as a protein synthesis inhibitor
  • Exhibits anti-HSV-1 activity
  • Belongs to the tetracycline class of antibiotics
  • Classified as a phenol