Learn More
Medchemexpress LLC Oxytetracycline dihydrate | 6153-64-6 | 99.4% | 500 MG
SDP
Catalog No. 5000408749
Encompass_Preferred
Oxytetracycline dihydrate is an antibiotic of the tetracycline class, acting as a potent inhibitor of both Gram-negative and Gram-positive bacteria. It functions by preventing the binding of aminoacyl-tRNA to the m-ribosomal RNA complex and also exhibits anti-HSV-1 activity. This compound is synthesized by a type II polyketide synthase from microorganisms, specifically *Streptomyces rimosus*.
  • Inhibits Gram-negative and Gram-positive bacteria
  • Acts as a protein synthesis inhibitor
  • Exhibits anti-HSV-1 activity
  • Light yellow to yellow solid appearance
Catalog No. 50-004-08749 Supplier Medchemexpress LLC Supplier No. HYB0275B500MG
Oxytetracycline dihydrate is an antibiotic of the tetracycline class, acting as a potent inhibitor of both Gram-negative and Gram-positive bacteria. It functions by preventing the binding of aminoacyl-tRNA to the m-ribosomal RNA complex and also exhibits anti-HSV-1 activity. This compound is synthesized by a type II polyketide synthase from microorganisms, specifically *Streptomyces rimosus*.
  • Inhibits Gram-negative and Gram-positive bacteria
  • Acts as a protein synthesis inhibitor
  • Exhibits anti-HSV-1 activity
  • Light yellow to yellow solid appearance