Learn More
Medchemexpress LLC Oxytetracycline dihydrate | 6153-64-6 | MFCD00151234 | 99.4% | 100 MG
SDP
Catalog No. 5000409019
Encompass_Preferred
Oxytetracycline dihydrate is an antibiotic of the tetracycline class that potently inhibits Gram-negative and Gram-positive bacteria. It acts as a protein synthesis inhibitor by preventing the binding of aminoacil-tRNA to the m-ribosomal RNA complex and also exhibits anti-HSV-1 activity.
  • Potently inhibits gram-negative and gram-positive bacteria
  • Acts as a protein synthesis inhibitor
  • Prevents the binding of aminoacil-tRNA to the m-ribosomal RNA complex
  • Exhibits anti-HSV-1 activity
  • Bacterial aromatic polyketide
Catalog No. 50-004-09019 Supplier Medchemexpress LLC Supplier No. HYB0275B100MG
Oxytetracycline dihydrate is an antibiotic of the tetracycline class that potently inhibits Gram-negative and Gram-positive bacteria. It acts as a protein synthesis inhibitor by preventing the binding of aminoacil-tRNA to the m-ribosomal RNA complex and also exhibits anti-HSV-1 activity.
  • Potently inhibits gram-negative and gram-positive bacteria
  • Acts as a protein synthesis inhibitor
  • Prevents the binding of aminoacil-tRNA to the m-ribosomal RNA complex
  • Exhibits anti-HSV-1 activity
  • Bacterial aromatic polyketide