Learn More
TARGETMOL CHEMICALS INC Oxytetracycline Dihydrate
SDP
Catalog No. 502880490
Encompass_Preferred
Also available in 1 g, 1 mL, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Oxytetracycline Dihydrate (Terramycin Dihydrate) is a prescription antibiotic, interfering with the ability of bacteria to produce essential proteins. Purity 97.3%
Catalog No. 50-288-0490 Supplier TARGETMOL CHEMICALS INC Supplier No. T6624200MG
Also available in 1 g, 1 mL, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Oxytetracycline Dihydrate (Terramycin Dihydrate) is a prescription antibiotic, interfering with the ability of bacteria to produce essential proteins. Purity 97.3%