Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Oxytetracycline Dihydrate, G-Biosciences
SCI_ED

Catalog No. S139808
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S139808 50 g
S000004 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S139808 Supplier G-Biosciences Supplier No. RC1079
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Oxytetracycline is a tetracycline antibiotic derived from Streptomyces rimosus. It targets gram-positive and gram-negative bacteria and several Mycoplasma species.

Oxytetracycline has been used to study its resistance gene, otrA and in fluorescence analysis of liver cells. Additionally, oxytetracycline is used as a selective agent in OGYE, a selective medium for yeasts and molds. It has been used to prevent fire blight in apples and pears.

Chemical Identifiers

CAS 6153-64-6
Molecular Weight (g/mol) 496.46 g/mol

Specifications

Technique HPLC, Spectrometry, UV/Vis, Ion Chromatography, Mass Spectrometry
Certifications/Compliance ISO, ISO 9001, REACH, GHS/OSHA, NIST
Type Drug Standard
Element Multi-element
Industry Type Biotech
Quantity 50 g
Percent Purity 97%
Grade Molecular Biology
Chemical Name or Material Oxytetracycline Dihydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.