Learn More
Research Products International Corp OXYTETRACYCLINE HCL, 5GM
SDP
Catalog No. 50213586
Encompass_Preferred
Oxytetracycline Hydrochloride is a tetracycline antibiotic.  Oxytetracycline inhibits bacterial protein synthesis by binding to the 30S ribosomal subunit, preventing the attachment of aminoacyl-tRNA to the mRNA-ribosome complex. The interference disrupts the translation process and inhibits bacterial growth. 
This antibiotic offers a broad spectrum of antibacterial activity against both Gram-positive and Gram-negative bacteria.  Additionally, it can be used to study bacterial susceptibility patterns or as a selective agent in culture media to isolate bacteria that are sensitive to this antibiotic.
WARNING! This product can expose you to Ocytetracycline HCl, a chemical which is known to the State of California to cause developmental harm. For more information go to www.P65Warnings.ca.gov.
Catalog No. 50-213-586 Supplier Research Products International Corp Supplier No. O200205.0
Oxytetracycline Hydrochloride is a tetracycline antibiotic.  Oxytetracycline inhibits bacterial protein synthesis by binding to the 30S ribosomal subunit, preventing the attachment of aminoacyl-tRNA to the mRNA-ribosome complex. The interference disrupts the translation process and inhibits bacterial growth. 
This antibiotic offers a broad spectrum of antibacterial activity against both Gram-positive and Gram-negative bacteria.  Additionally, it can be used to study bacterial susceptibility patterns or as a selective agent in culture media to isolate bacteria that are sensitive to this antibiotic.
WARNING! This product can expose you to Ocytetracycline HCl, a chemical which is known to the State of California to cause developmental harm. For more information go to www.P65Warnings.ca.gov.