Learn More
Medchemexpress LLC Oxytetracycline hydrochloride | 2058-46-0 | 98.57% | 50 MG
SDP
Catalog No. 5000406601
Encompass_Preferred
Oxytetracycline hydrochloride is an analytical standard of Oxytetracycline (hydrochloride) intended for research and analytical applications. It is an antibiotic from the tetracycline class, known for potently inhibiting both Gram-negative and Gram-positive bacteria. This compound acts as a protein synthesis inhibitor and also exhibits anti-HSV-1 activity.
  • Potently inhibits Gram-negative and Gram-positive bacteria
  • Functions as a protein synthesis inhibitor
  • Prevents binding of aminoacyl-tRNA to the m-ribosomal RNA complex
  • Exhibits anti-HSV-1 activity
  • Used as an analytical standard for qualitative, quantitative, and methodological research experiments
  • Suitable for techniques such as HPLC, GC, and MS
Catalog No. 50-004-06601 Supplier Medchemexpress LLC Supplier No. HYB0275AR50MG
Oxytetracycline hydrochloride is an analytical standard of Oxytetracycline (hydrochloride) intended for research and analytical applications. It is an antibiotic from the tetracycline class, known for potently inhibiting both Gram-negative and Gram-positive bacteria. This compound acts as a protein synthesis inhibitor and also exhibits anti-HSV-1 activity.
  • Potently inhibits Gram-negative and Gram-positive bacteria
  • Functions as a protein synthesis inhibitor
  • Prevents binding of aminoacyl-tRNA to the m-ribosomal RNA complex
  • Exhibits anti-HSV-1 activity
  • Used as an analytical standard for qualitative, quantitative, and methodological research experiments
  • Suitable for techniques such as HPLC, GC, and MS