Learn More
Medchemexpress LLC Oxytetracycline hydrochloride | 2058-46-0 | 98.9% | 10 G
SDP
Catalog No. 5000406450
Encompass_Preferred
Oxytetracycline hydrochloride is an antibiotic from the tetracycline class that potently inhibits Gram-negative and Gram-positive bacteria. It functions as a protein synthesis inhibitor, preventing the binding of aminoacyl-tRNA to the m-ribosomal RNA complex. Additionally, it exhibits anti-HSV-1 activity.
  • Antibiotic from the tetracycline class
  • Potently inhibits Gram-negative and Gram-positive bacteria
  • Functions as a protein synthesis inhibitor
  • Exhibits anti-HSV-1 activity
Catalog No. 50-004-06450 Supplier Medchemexpress LLC Supplier No. HYB0275A10G
Oxytetracycline hydrochloride is an antibiotic from the tetracycline class that potently inhibits Gram-negative and Gram-positive bacteria. It functions as a protein synthesis inhibitor, preventing the binding of aminoacyl-tRNA to the m-ribosomal RNA complex. Additionally, it exhibits anti-HSV-1 activity.
  • Antibiotic from the tetracycline class
  • Potently inhibits Gram-negative and Gram-positive bacteria
  • Functions as a protein synthesis inhibitor
  • Exhibits anti-HSV-1 activity