Learn More
Medchemexpress LLC Oxytetracycline (hydrochloride) | 2058-46-0 | 98.9% | 25 MG
SDP
Catalog No. 5000450996
Encompass_Preferred
Oxytetracycline hydrochloride is an antibiotic belonging to the tetracycline class, known for its potent inhibition of Gram-negative and Gram-positive bacteria. This compound acts as a protein synthesis inhibitor, preventing the binding of aminoacyl-tRNA to the m-ribosomal RNA complex. It also exhibits anti-HSV-1 activity and is synthesized by a type II polyketide synthase. This product is for research use only.
  • Potently inhibits Gram-negative bacteria
  • Potently inhibits Gram-positive bacteria
  • Functions as a protein synthesis inhibitor
  • Prevents aminoacyl-tRNA binding to m-ribosomal RNA complex
  • Exhibits anti-HSV-1 activity
  • Stable under recommended storage conditions
  • Appears as a light yellow to yellow solid
Catalog No. 50-004-50996 Supplier Medchemexpress LLC Supplier No. 25MGHYB0275AR
Oxytetracycline hydrochloride is an antibiotic belonging to the tetracycline class, known for its potent inhibition of Gram-negative and Gram-positive bacteria. This compound acts as a protein synthesis inhibitor, preventing the binding of aminoacyl-tRNA to the m-ribosomal RNA complex. It also exhibits anti-HSV-1 activity and is synthesized by a type II polyketide synthase. This product is for research use only.
  • Potently inhibits Gram-negative bacteria
  • Potently inhibits Gram-positive bacteria
  • Functions as a protein synthesis inhibitor
  • Prevents aminoacyl-tRNA binding to m-ribosomal RNA complex
  • Exhibits anti-HSV-1 activity
  • Stable under recommended storage conditions
  • Appears as a light yellow to yellow solid