Learn More
Medchemexpress LLC Oxytetracycline hydrochloride | 2058-46-0 | 98.9% | 5 G
SDP
Catalog No. 5000406075
Encompass_Preferred
Oxytetracycline hydrochloride is an antibiotic from the tetracycline class. It effectively inhibits both Gram-negative and Gram-positive bacteria, acting as a protein synthesis inhibitor by preventing aminoacyl-tRNA binding to the ribosomal RNA complex. This compound also exhibits anti-HSV-1 activity.
  • Antibiotic from the tetracycline class
  • Potently inhibits Gram-negative and Gram-positive bacteria
  • Protein synthesis inhibitor
  • Prevents aminoacyl-tRNA binding to the ribosomal RNA complex
  • Exhibits anti-HSV-1 activity
Catalog No. 50-004-06075 Supplier Medchemexpress LLC Supplier No. HYB0275A5G
Oxytetracycline hydrochloride is an antibiotic from the tetracycline class. It effectively inhibits both Gram-negative and Gram-positive bacteria, acting as a protein synthesis inhibitor by preventing aminoacyl-tRNA binding to the ribosomal RNA complex. This compound also exhibits anti-HSV-1 activity.
  • Antibiotic from the tetracycline class
  • Potently inhibits Gram-negative and Gram-positive bacteria
  • Protein synthesis inhibitor
  • Prevents aminoacyl-tRNA binding to the ribosomal RNA complex
  • Exhibits anti-HSV-1 activity