Learn More
Medchemexpress LLC Oxytetracycline hydrochloride | 2058-46-0 | MFCD00135815 | 98.92% | 1 ML
SDP
Catalog No. 5000406040
Encompass_Preferred
Oxytetracycline hydrochloride is an antibiotic belonging to the tetracycline class. It potently inhibits Gram-negative and Gram-positive bacteria. It acts as a protein synthesis inhibitor, preventing the binding of aminoacyl-tRNA to the m-ribosomal RNA complex. It also possesses anti-HSV-1 activity.
  • Potently inhibits Gram-negative and Gram-positive bacteria
  • Acts as a protein synthesis inhibitor
  • Exhibits anti-HSV-1 activity
Catalog No. 50-004-06040 Supplier Medchemexpress LLC Supplier No. HYB027510MM/1ML
Oxytetracycline hydrochloride is an antibiotic belonging to the tetracycline class. It potently inhibits Gram-negative and Gram-positive bacteria. It acts as a protein synthesis inhibitor, preventing the binding of aminoacyl-tRNA to the m-ribosomal RNA complex. It also possesses anti-HSV-1 activity.
  • Potently inhibits Gram-negative and Gram-positive bacteria
  • Acts as a protein synthesis inhibitor
  • Exhibits anti-HSV-1 activity