Learn More
Alkali Scientific Oxytetracycline Hydrochloride, 5 G
SDP
Catalog No. 5000660110
Encompass_Preferred
Oxytetracycline Hydrochloride, available in a 5g laboratory-grade powder form, is a broad-spectrum antibiotic effective in treating a wide variety of bacterial infections. This compound works by inhibiting protein synthesis in bacteria, making it an essential tool in antimicrobial research. It is commonly used for testing bacterial susceptibility in clinical microbiology, particularly for organisms like Staphylococcus and Escherichia coli. In addition to medical applications, oxytetracycline is used in veterinary medicine and agriculture. The 5g powder format is ideal for preparing precise concentrations for use in bacteriological media or in solution for antimicrobial testing.
Catalog No. 50-006-60110 Supplier Alkali Scientific Supplier No. OXY002CP
Oxytetracycline Hydrochloride, available in a 5g laboratory-grade powder form, is a broad-spectrum antibiotic effective in treating a wide variety of bacterial infections. This compound works by inhibiting protein synthesis in bacteria, making it an essential tool in antimicrobial research. It is commonly used for testing bacterial susceptibility in clinical microbiology, particularly for organisms like Staphylococcus and Escherichia coli. In addition to medical applications, oxytetracycline is used in veterinary medicine and agriculture. The 5g powder format is ideal for preparing precise concentrations for use in bacteriological media or in solution for antimicrobial testing.