Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Oxytetracycline hydrochloride, can be used as secondary standard'

Catalog No. AC301275000
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC301275000 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC301275000 Supplier Thermo Scientific Chemicals Supplier No. 301275000
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2058-46-0
Molecular Formula C22H25ClN2O9
Molecular Weight (g/mol) 496.897
MDL Number MFCD00135815
InChI Key UTAHTGPLLFLKGW-IFLJXUKPSA-N
Synonym 5-Hydroxytetracycline hydrochloride
PubChem CID 121235537
IUPAC Name (4S,4aR,5S,5aR,6S,12aS)-4-(dimethylamino)-3,5,6,10,11,12a-hexahydroxy-6-methyl-1,12-dioxo-4,4a,5,5a-tetrahydrotetracene-2-carboxamide;hydrochloride
SMILES CC1(C2C(C3C(C(=C(C(=O)C3(C(=O)C2=C(C4=C1C=CC=C4O)O)O)C(=O)N)O)N(C)C)O)O.Cl

Specifications

Type Oxytetracycline Hydrochloride
Physical Form Crystalline Powder
Infrared Spectrum Authentic
Packaging Glass Bottle
Color Beige to Yellow
Quantity 500 mg
Beilstein 14, IV, 2633
Merck Index 15, 7075
Solubility Information Solubility in water: >100g/L. Other solubilities: 20g/L methanol
Formula Weight 496.9
Percent Purity ≥97.0% Total Base (anhydrous and solvent free), ≥80 wt% (as is, calculated, HPLC)
Chemical Name or Material Oxytetracycline hydrochloride
Show More Show Less
Hazard Category LIGHT SENSITIVE
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Suspected of damaging the unborn child.
Precautionary Statement GHS P Statement:
Use personal protective equipment as required.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.