Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Oxytetracycline Hydrochloride 95.0+%, TCI America™
SDP

Catalog No. O047525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O047525G 25 g
O04755G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O047525G Supplier TCI America Supplier No. O047525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2058-46-0
Molecular Formula C22H25ClN2O9
Molecular Weight (g/mol) 496.897
MDL Number MFCD00135815
InChI Key UTAHTGPLLFLKGW-IFLJXUKPSA-N
PubChem CID 121235537
IUPAC Name (4S,4aR,5S,5aR,6S,12aS)-4-(dimethylamino)-3,5,6,10,11,12a-hexahydroxy-6-methyl-1,12-dioxo-4,4a,5,5a-tetrahydrotetracene-2-carboxamide;hydrochloride
SMILES CC1(C2C(C3C(C(=C(C(=O)C3(C(=O)C2=C(C4=C1C=CC=C4O)O)O)C(=O)N)O)N(C)C)O)O.Cl

Specifications

Melting Point 204°C
Color Yellow
Quantity 25 g
UN Number 3077
Formula Weight 496.90
Percent Purity ≥95.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Oxytetracycline Hydrochloride
EINECSNumber (9)-0271
RTECSNumber QI8225000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.