Learn More
Apexbio Technology LLC Oxytetracycline (Terramycin) 79-57-2 10mM (in 1mL DMSO)
SDP
Catalog No. 5000092885
Encompass_Preferred
Oxytetracycline (Terramycin) (CAS 79-57-2) is a small-molecule inhibitor targeting bacterial protein synthesis It is designed to inhibit aminoacyl-tRNA binding to the 30S ribosomal subunit thereby interfering with bacterial protein synthesis Oxytetracycline exerts its biological activity primarily through reversible binding to the bacterial 30S ribosomal subunit inhibiting protein biosynthesis In in vitro studies Oxytetracycline demonstrates bacteriostatic activity against Gram-positive and Gram-negative bacteria with an IC50 value typically ranging between 0 1 10 g/mL depending on the tested microorganism and experimental system Based on these pharmacological properties Oxytetracycline holds research potential in microbiological studies to suppress contaminating bacteria and in cell culture research for maintaining sterile conditions or selecting antibiotic-resistant cell populations
Catalog No. 50-000-92885 Supplier Apexbio Technology LLC Supplier No. B20005.1
Oxytetracycline (Terramycin) (CAS 79-57-2) is a small-molecule inhibitor targeting bacterial protein synthesis It is designed to inhibit aminoacyl-tRNA binding to the 30S ribosomal subunit thereby interfering with bacterial protein synthesis Oxytetracycline exerts its biological activity primarily through reversible binding to the bacterial 30S ribosomal subunit inhibiting protein biosynthesis In in vitro studies Oxytetracycline demonstrates bacteriostatic activity against Gram-positive and Gram-negative bacteria with an IC50 value typically ranging between 0 1 10 g/mL depending on the tested microorganism and experimental system Based on these pharmacological properties Oxytetracycline holds research potential in microbiological studies to suppress contaminating bacteria and in cell culture research for maintaining sterile conditions or selecting antibiotic-resistant cell populations