Learn More
Selleck Chemical LLC Oxythiamine HCl-E1800-250MG
SDP
Catalog No. 503138945
Encompass_Preferred
Oxythiamine chloride hydrochloride(Hydroxythiamine chloride hydrochloride) is a salt form of Oxythiamine It is an inhibitor of transketolase the enzyme that controls the nonoxidative branch of the pentose phosphate pathway It also suppresses tumor cell proliferation and promotes apoptosis
Catalog No. 50-313-8945 Supplier Selleck Chemical LLC Supplier No. E1800250MG
Oxythiamine chloride hydrochloride(Hydroxythiamine chloride hydrochloride) is a salt form of Oxythiamine It is an inhibitor of transketolase the enzyme that controls the nonoxidative branch of the pentose phosphate pathway It also suppresses tumor cell proliferation and promotes apoptosis