Learn More
CPC Scientific H-Cys-Tyr-Ile-Gln-Asn-Cys-Pro-Leu-Gly-NH2 (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific OXYT001A
Encompass
ONE-LETTER SEQUENCE: CYIQNCPLG-NH2 (C1&C6 bridge)MOLECULAR FORMULA: C43H66N12O12S2MOLECULAR WEIGHT:1007.2STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [50-56-6], RESEARCH AREA: HormonalREFERENCES: M.Nagasawa et al., Science, 348, 333 (2015); H.-J.Lee et al., Progr. Neurobiol., 88, 127 (2009); I.D.Neumann, J. Neuroendocrinol., 20, 858 (2008); J.G.Smith and D.C.Merrill, Clin. Obstet. Gynecol., 49, 594 (2006); T.H.Lippert et al., Horm. Res., 60, 262 (2003)
Catalog No. 50-211-0045