Learn More
TARGETMOL CHEMICALS INC Oxytocin acetate 50MG
SDP
Catalog No. 502879783
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 25 mg, 100 mg and bulk. Please contact Fisher for quotes. Oxytocin acetate (alpha-Hypophamine acetate) is a mammalian neurohypophysial hormone, a ligand of oxytocin receptor.
Catalog No. 50-287-9783 Supplier TARGETMOL CHEMICALS INC Supplier No. T514550MG
Also available in 1 mL, 5 mg, 10 mg, 25 mg, 100 mg and bulk. Please contact Fisher for quotes. Oxytocin acetate (alpha-Hypophamine acetate) is a mammalian neurohypophysial hormone, a ligand of oxytocin receptor.