Learn More
Medchemexpress LLC Oxytocin acetate | 6233-83-6 | 99.66% | 1067.2 g/mol | 5 MG
SDP
Catalog No. 5000393390
Encompass_Preferred
Oxytocin (α-Hypophamine) acetate is a pleiotropic, hypothalamic peptide known for facilitating parturition, lactation, and prosocial behaviors. It can function as a stress-coping molecule with anti-inflammatory, antioxidant, and protective effects, especially in the face of adversity or trauma.
  • Known for facilitating parturition, lactation, and prosocial behaviors
  • Functions as a stress-coping molecule
  • Offers anti-inflammatory, antioxidant, and protective effects
Catalog No. 50-003-93390 Supplier Medchemexpress LLC Supplier No. 5MGHY17571A
Oxytocin (α-Hypophamine) acetate is a pleiotropic, hypothalamic peptide known for facilitating parturition, lactation, and prosocial behaviors. It can function as a stress-coping molecule with anti-inflammatory, antioxidant, and protective effects, especially in the face of adversity or trauma.
  • Known for facilitating parturition, lactation, and prosocial behaviors
  • Functions as a stress-coping molecule
  • Offers anti-inflammatory, antioxidant, and protective effects