Learn More
Medchemexpress LLC Oxytocin (acetate) | 6233-83-6 | 99.7% | 1067.24 | 100 MG
SDP
Catalog No. 5000387458
Encompass_Preferred
Oxytocin acetate is a pleiotropic, hypothalamic peptide known for facilitating parturition, lactation, and prosocial behaviors. It functions as a stress-coping molecule with anti-inflammatory, antioxidant, and protective effects, especially in the face of adversity or trauma.
  • Pleiotropic hypothalamic peptide
  • Facilitates parturition, lactation, and prosocial behaviors
  • Functions as a stress-coping molecule
  • Exhibits anti-inflammatory effects
  • Acts as an antioxidant
  • Provides protective effects, particularly during adversity or trauma
Catalog No. 50-003-87458 Supplier Medchemexpress LLC Supplier No. HY17571A100MG
Oxytocin acetate is a pleiotropic, hypothalamic peptide known for facilitating parturition, lactation, and prosocial behaviors. It functions as a stress-coping molecule with anti-inflammatory, antioxidant, and protective effects, especially in the face of adversity or trauma.
  • Pleiotropic hypothalamic peptide
  • Facilitates parturition, lactation, and prosocial behaviors
  • Functions as a stress-coping molecule
  • Exhibits anti-inflammatory effects
  • Acts as an antioxidant
  • Provides protective effects, particularly during adversity or trauma