Learn More
Medchemexpress LLC Oxytocin (acetate) | 6233-83-6 | 99.7% | 1067.24 | 50 MG
SDP
Catalog No. 5000386918
Encompass_Preferred
Oxytocin (α-Hypophamine) acetate is a pleiotropic, hypothalamic peptide known for facilitating parturition, lactation, and prosocial behaviors. It can also function as a stress-coping molecule with anti-inflammatory, antioxidant, and protective effects, particularly in situations of adversity or trauma.
  • Facilitates parturition and lactation.
  • Promotes prosocial behaviors.
  • Acts as a stress-coping molecule.
  • Exhibits anti-inflammatory effects.
  • Provides antioxidant benefits.
  • Offers protective effects, especially in the face of adversity or trauma.
Catalog No. 50-003-86918 Supplier Medchemexpress LLC Supplier No. HY17571A50MG
Oxytocin (α-Hypophamine) acetate is a pleiotropic, hypothalamic peptide known for facilitating parturition, lactation, and prosocial behaviors. It can also function as a stress-coping molecule with anti-inflammatory, antioxidant, and protective effects, particularly in situations of adversity or trauma.
  • Facilitates parturition and lactation.
  • Promotes prosocial behaviors.
  • Acts as a stress-coping molecule.
  • Exhibits anti-inflammatory effects.
  • Provides antioxidant benefits.
  • Offers protective effects, especially in the face of adversity or trauma.