Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Oxytocin acetate | 6233-83-6 | MFCD11046045 | 99.7% | 10 MG
SDP

Catalog No. 5000298398
Encompass_Preferred

Oxytocin acetate is an endogenous hypothalamic nonapeptide supplied as the acetate salt for research use as an oxytocin receptor agonist. Provided as a powdered reagent, it is used in biochemical, pharmacological, and in vivo studies to probe oxytocin signaling and physiological responses.

  • High purity, ~99.7%, appropriate for biochemical assays.
  • Nonapeptide sequence: Cys-Tyr-Ile-Gln-Asn-Cys-Pro-Leu-Gly-NH2 (disulfide bridge Cys1-Cys6).
  • Soluble in DMSO at 125 mg/mL; in vivo formulation achievable at ≥2.08 mg/mL.
  • Stable as powder when stored sealed at -80°C (2 years) or -20°C (1 year).
  • Suitable as a reagent or reference standard for receptor and signaling studies.

Catalog No. 50-002-98398 Supplier Medchemexpress LLC Supplier No. HY17571A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Oxytocin acetate is an endogenous hypothalamic nonapeptide supplied as the acetate salt for research use as an oxytocin receptor agonist. Provided as a powdered reagent, it is used in biochemical, pharmacological, and in vivo studies to probe oxytocin signaling and physiological responses.

  • High purity, ~99.7%, appropriate for biochemical assays.
  • Nonapeptide sequence: Cys-Tyr-Ile-Gln-Asn-Cys-Pro-Leu-Gly-NH2 (disulfide bridge Cys1-Cys6).
  • Soluble in DMSO at 125 mg/mL; in vivo formulation achievable at ≥2.08 mg/mL.
  • Stable as powder when stored sealed at -80°C (2 years) or -20°C (1 year).
  • Suitable as a reagent or reference standard for receptor and signaling studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.