Learn More
Cayman Chemical OxytocIn AChE Tracer 500dtn
SDP
Supplier:  Cayman Chemical 400440500 DTN
Encompass_Preferred
A prodrug form of 8-bromo-cADPR prevents fMLP-induced increases in intracellular calcium levels and chemotaxis in isolated mouse bone marrow-derived neutrophils at 1 mM decreases LPS-induced nitrite production and TNF-a and IL-2 secretion in mouse primary microglial cells at 100 UM
Catalog No. 50-294-2481