Learn More
Enzo Life Sciences Oxytocin ELISA kit (5x96wells)
SDP
Supplier:  Enzo Life Sciences ADI901153A0001
Encompass_Preferred
The most sensitive, specific and flexible alternative to more costly and time-consuming methods for oxytocin detection. The Oxytocin EIA kit is a colorimetric competitive enzyme immunoassay kit with results overnight + 1 hour. Absorbance is read at 405 nm. Very low reactivity with vasopressin provides confidence in assay results. Sensitive quantification of 15pg/mL oxytocin. Ready-to-use liquid color-coded reagents reduce errors in the lab. Sensitivity: 15pg/ml (range 15.6 - 1,000 pg/ml). Assay Time: Overnight + 1 hour. Applications: ELISA, Colorimetric detection. Species Reactivity: Species independent. Contents: GxR Microtiter plate, Conjugate, Antibody, Assay buffer concentrate, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution
Catalog No. NC0733090