Learn More
Arbor Assays Oxytocin ELISA Kit (1 Strip Plate)
SDP
Supplier:  Arbor Assays K048H1
Encompass_Preferred
The DetectX Oxytocin ELISA Kit is designed to quantitatively measure Oxytocin present in serum, plasma, clarified milk and tissue culture media samples.
Catalog No. 50-227-8459