Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Oxytocin parallel dimer TFA | 19645-28-4 | 98.1% | 2128.40 | 1 MG
SDP

Catalog No. 5000398730
Encompass_Preferred

Oxytocin parallel dimer TFA is a disulfide-bridged homo peptide dimer. It exhibits oxytocin and vasopressin-like activity with reduced toxicity compared to oxytocin and is intended for research use only.

  • Disulfide-bridged homo peptide dimer
  • Exhibits oxytocin and vasopressin-like activity
  • Less toxic than oxytocin
  • For research use only
  • Purity: 98.1%
  • Molecular weight: 2128.40
  • Appearance: White to off-white solid
  • Appropriate storage conditions for powder and in solvent

Catalog No. 50-003-98730 Supplier Medchemexpress LLC Supplier No. HYP3215A1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Oxytocin parallel dimer TFA is a disulfide-bridged homo peptide dimer. It exhibits oxytocin and vasopressin-like activity with reduced toxicity compared to oxytocin and is intended for research use only.

  • Disulfide-bridged homo peptide dimer
  • Exhibits oxytocin and vasopressin-like activity
  • Less toxic than oxytocin
  • For research use only
  • Purity: 98.1%
  • Molecular weight: 2128.40
  • Appearance: White to off-white solid
  • Appropriate storage conditions for powder and in solvent

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.